ADV412110371 AChemBlock

4-methyl-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde

  • MDL number MFCD11616739
  • Cas Number : 4894-34-2
  • Molecular Weight : 160.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1h-pyrrolo-2-3-b-pyridine-3-carboxaldehyde-4-methyl.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4718
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart