ADV412110376 AChemBlock

2,6-Diazaspiro[3.3]heptane dihydrochloride

  • MDL number MFCD11878485
  • Cas Number : 1184963-68-5
  • Molecular Weight : 171.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-6-diazaspiro-3-3-heptane-dihydrochloride.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4724
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart