ADV412110403 AChemBlock

(2-Chloro-5-methyl-pyrimidin-4-yl)-(3-nitro-phenyl)-amine

  • MDL number MFCD26385095
  • Cas Number : 1202759-74-7
  • Molecular Weight : 264.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-5-methyl-pyrimidin-4-yl-3-nitro-phenyl-amine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4753
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart