ADV412110408 AChemBlock

Spiro[indole-3,3`-pyrrolidin]-2(1H)-one

  • MDL number MFCD17019392
  • Cas Number : 6786-41-0
  • Molecular Weight : 188.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Physical Form white solid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/spiro-indole-3-3-pyrrolidin-2-1h-one.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4758
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart