ADV412110410 AChemBlock

2-Oxa-6-azaspiro[3.4]octan-7-one

  • MDL number MFCD14586414
  • Cas Number : 1207174-87-5
  • Molecular Weight : 127.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-oxa-6-azaspiro-3-4-octan-7-one.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4760
pictograms GHS07
hstatements H303;H335
Availability details for
Quantity
  • Availability
  • Add to Cart