ADV412110414 AChemBlock

3-Oxabicyclo[3.1.0]hexane-6-carboxylic acid

  • MDL number MFCD13193581
  • Cas Number : 693248-53-2
  • Molecular Weight : 128.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-oxabicyclo-3-1-0-hexane-6-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4765
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart