ADV412110418 AChemBlock

(+/-)-Cbz-alpha-phosphonoglycine trimethyl ester

  • MDL number MFCD00043304
  • Cas Number : 88568-95-0
  • Molecular Weight : 331.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/cbz-alpha-phosphonoglycine-trimethyl-ester.html

Safety Information

signalword Danger
pcodes P261;P301+310;P305+351+338;P361
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3344
pictograms GHS06; GHS07
hstatements H301;H311;H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart