ADV412110421 AChemBlock

6-Chloro-2-nitropyridin-3-ol

  • MDL number MFCD09834122
  • Cas Number : 887471-39-8
  • Molecular Weight : 174.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-chloro-2-nitropyridin-3-ol.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4775
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart