ADV412110422 AChemBlock

Cyclopropylmethanethiol

  • MDL number MFCD00051613
  • Cas Number : 5617-79-8
  • Molecular Weight : 88.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/cyclopropylmethanethiol.html

Safety Information

signalword Danger
pcodes P280;P310;P261;P305+351+338;P302+350;P301+330+331
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4776
pictograms GHS05;GHS06;GHS07
hstatements H226;H302;H311;H314;H335
Availability details for
Quantity
  • Availability
  • Add to Cart