ADV412110437 AChemBlock

(2-Amino-thiazol-5-yl)-acetic acid methyl ester hydrobromide

  • MDL number MFCD18432774
  • Cas Number : 723278-34-0
  • Molecular Weight : 253.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-amino-thiazol-5-yl-acetic-acid-methyl-ester-hydrobromide.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3490
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart