ADV412110438 AChemBlock

(2S)-1-(tert-Butoxycarbonyl)-2-methyl-2-pyrrolidinecarboxylic acid

  • MDL number MFCD08706361
  • Cas Number : 103336-06-7
  • Molecular Weight : 229.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2s-1-tert-butoxycarbonyl-2-methyl-2-pyrrolidinecarboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4786
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart