ADV412110458 AChemBlock

MLN9708

  • MDL number MFCD18251437
  • Cas Number : 1201902-80-8
  • Molecular Weight : 517.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/mln9708.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4808
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart