ADV412110462 AChemBlock

3-Amino-cyclobutanemethanol hydrochloride

  • MDL number MFCD16659926
  • Cas Number : 130369-06-1
  • Molecular Weight : 137.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow liquid/orange liquid/colorless liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-amino-cyclobutanemethanol-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3529
pictograms GHS07
hstatements H303
Availability details for
Quantity
  • Availability
  • Add to Cart