ADV412110473 AChemBlock

5-Aminomethyl-7-azaindole hydrochloride

  • MDL number MFCD19382526
  • Cas Number : 1432754-52-3
  • Molecular Weight : 183.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-aminomethyl-7-azaindole-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3530
pictograms GHS07
hstatements H303
Availability details for
Quantity
  • Availability
  • Add to Cart