ADV412110485 AChemBlock

4-(2-Nitroethenyl)pyridine

  • MDL number MFCD11878108
  • Cas Number : 100446-37-5
  • Molecular Weight : 150.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form brown solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/4-2-nitroethenyl-pyridine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4841
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart