ADV412110486 AChemBlock

1-(Aminomethyl)cyclopropanol

  • MDL number MFCD04972522
  • Cas Number : 74592-33-9
  • Molecular Weight : 87.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-aminomethyl-cyclopropanol.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4842
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart