ADV412110489 AChemBlock

1-Methyl-4-(2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)ethyl)piperazine

  • MDL number MFCD22378010
  • Cas Number : 1392419-83-8
  • Molecular Weight : 320.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow liquid/white solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-methyl-4-2-4-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-1h-pyrazol-1-yl-ethyl-piperazine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4845
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart