ADV412110496 AChemBlock

tert-Butyl (Morpholin-3-ylmethyl)carbamate

  • MDL number MFCD11111574
  • Cas Number : 169750-75-8
  • Molecular Weight : 216.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-morpholin-3-ylmethyl-carbamate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4853
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart