ADV412110504 AChemBlock

1-Azetidin-3-ylpyrrolidine dihydrochloride

  • MDL number MFCD11617821
  • Cas Number : 1024589-68-1
  • Molecular Weight : 199.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form white powder
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-azetidin-3-ylpyrrolidine-dihydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4871
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart