ADV412110515 AChemBlock

5-Bromoisoindoline hydrochloride

  • MDL number MFCD10700135
  • Cas Number : 919346-89-7
  • Molecular Weight : 234.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form gray solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-bromoisoindoline-hydrochloride.html

Safety Information

signalword Danger
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3625
pictograms GHS05; GHS07
hstatements H302;H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart