ADV412110531 AChemBlock

cis-3-Amino-cyclobutanemethanol hydrochloride

  • MDL number MFCD16660356
  • Cas Number : 142733-65-1
  • Molecular Weight : 137.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid/yellow solid/off-white solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/cis-3-amino-cyclobutanemethanol-hydrochloride.html

Safety Information

signalword Warning
pcodes P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3635
pictograms GHS07
hstatements H303;H319
Availability details for
Quantity
  • Availability
  • Add to Cart