ADV412110532 AChemBlock

trans-3-Amino-cyclobutanemethanol hydrochloride

  • MDL number MFCD22628707
  • Cas Number : 1284250-10-7
  • Molecular Weight : 137.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form off-white solid/light-yellow liquid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/trans-3-amino-cyclobutanemethanol-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P264;P270;P271;P280;P301+312;P302+352;P304+340;P322;P363
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3636
pictograms GHS07
hstatements H302;H312;H332
Availability details for
Quantity
  • Availability
  • Add to Cart