ADV412110534 AChemBlock

Exo-3-oxabicyclo[3.1.0]hexane-6-carboxylic acid

  • MDL number MFCD19228486
  • Cas Number : 55780-88-6
  • Molecular Weight : 128.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/exo-3-oxabicyclo-3-1-0-hexane-6-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8003
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart