ADV412110538 AChemBlock

(R)-tert-Butyl 1-aminopropan-2-ylcarbamate

  • MDL number MFCD11112229
  • Cas Number : 100927-10-4
  • Molecular Weight : 174.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/r-tert-butyl-1-aminopropan-2-ylcarbamate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8009
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart