ADV412110545 AChemBlock

Ethyl 3-amino-1-Boc-azetidine-3-carboxylate

  • MDL number MFCD18207531
  • Cas Number : 1011479-72-3
  • Molecular Weight : 244.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/ethyl-3-amino-1-boc-azetidine-3-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8016
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart