ADV412110556 AChemBlock

4-(Boc-amino)-3,6-dichloropyridazine

  • MDL number MFCD17214409
  • Cas Number : 887310-61-4
  • Molecular Weight : 264.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-boc-amino-3-6-dichloropyridazine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6735
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart