ADV412110584 AChemBlock

Methyl 5-bromo-2-oxo-1,2-dihydro-3-pyridinecarboxylate

  • MDL number MFCD03844843
  • Cas Number : 120034-05-1
  • Molecular Weight : 232.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Off white powder
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-5-bromo-2-oxo-1-2-dihydro-3-pyridinecarboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6765
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart