ADV412110590 AChemBlock

8-Bromoimidazo[1,2-a]pyridine hydrochloride

  • MDL number MFCD23106467
  • Cas Number : 1419101-42-0
  • Molecular Weight : 233.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/8-bromoimidazo-1-2-a-pyridine-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6771
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart