ADV412110592 AChemBlock

3-Pyrrolidineacetic acid

  • MDL number MFCD01318700
  • Cas Number : 89203-64-5
  • Molecular Weight : 129.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-pyrrolidineacetic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6773
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart