ADV412110597 AChemBlock

tert-Butyl 4-(2-bromoacetyl)piperidine-1-carboxylate

  • MDL number MFCD11975555
  • Cas Number : 301221-79-4
  • Molecular Weight : 306.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form rownish solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4-2-bromoacetyl-piperidine-1-carboxylate.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6778
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart