ADV412110602 AChemBlock

4-Bromo-2-methyl-1-phenylsulfonyl-7-azaindole

  • MDL number MFCD13191645
  • Cas Number : 1014613-05-8
  • Molecular Weight : 351.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-bromo-2-methyl-1-phenylsulfonyl-7-azaindole.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6783
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart