ADV412110610 AChemBlock

4-Nitro-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD00464007
  • Cas Number : 5334-40-7
  • Molecular Weight : 157.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form off-white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-nitro-1h-pyrazole-5-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P264;P271;P280;P302+352;P304+340;P305+351+338;P337+313;P403+233
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6797
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart