ADV412110622 AChemBlock

4-Chloro-6-fluoro-1,7-naphthyridine-3-carbonitrile

  • MDL number MFCD11109751
  • Cas Number : 305371-18-0
  • Molecular Weight : 207.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-chloro-6-fluoro-1-7-naphthyridine-3-carbonitrile.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6809
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart