ADV412110623 AChemBlock

3-Amino-4-bromo-1H-indazole

  • MDL number MFCD09909580
  • Cas Number : 914311-50-5
  • Molecular Weight : 212.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form pale cream powder
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-amino-4-bromo-1h-indazole.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6811
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart