ADV412110628 AChemBlock

3-Iodo-4-chloro-1-(phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridine

  • MDL number MFCD12923117
  • Cas Number : 1203566-61-3
  • Molecular Weight : 418.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-iodo-4-chloro-1-phenylsulfonyl-1h-pyrrolo-2-3-b-pyridine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6816
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart