ADV412110634 AChemBlock

4-Amino-2-chloro-5-iodopyridine

  • MDL number MFCD09834661
  • Cas Number : 800402-12-4
  • Molecular Weight : 254.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white to yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-amino-2-chloro-5-iodopyridine.html

Safety Information

signalword Danger
pcodes P264;P270;P280;P301+312;P305+351+338;P310
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6822
pictograms GHS05; GHS07
hstatements H302;H318
Availability details for
Quantity
  • Availability
  • Add to Cart