ADV412110637 AChemBlock

(3,4)-cis-tert-Butyl 3-amino-4-fluoropiperidine-1-carboxylate

  • MDL number MFCD18791212
  • Cas Number : 1273565-65-3
  • Molecular Weight : 218.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Light yellow liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-4-cis-tert-butyl-3-amino-4-fluoropiperidine-1-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6825
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart