ADV412110639 AChemBlock

Bicyclo[1.1.1]pentane-1,3-dicarboxylic acid

  • MDL number MFCD20621127
  • Cas Number : 56842-95-6
  • Molecular Weight : 156.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/bicyclo-1-1-1-pentane-1-3-dicarboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6827
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart