ADV412110656 AChemBlock

7-Boc-2-oxo-7-azabicyclo[2.2.1]heptane

  • MDL number MFCD18633018
  • Cas Number : 152533-47-6
  • Molecular Weight : 211.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white powder
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/7-boc-2-oxo-7-azabicyclo-2-2-1-heptane.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6846
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart