ADV412110661 AChemBlock

Gemigliptin

  • MDL number MFCD19443742
  • Cas Number : 911637-19-9
  • Molecular Weight : 489.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/gemigliptin.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6851
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart