ADV412110666 AChemBlock

4-Chloro-1-(triisopropylsilyl)-1H-pyrrolo[2,3-b]pyridin-5-ol

  • MDL number MFCD11616410
  • Cas Number : 685513-98-8
  • Molecular Weight : 324.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-chloro-1-triisopropylsilyl-1h-pyrrolo-2-3-b-pyridin-5-ol.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6856
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart