ADV412110679 AChemBlock

3-Chloro-1-methyl-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD19687914
  • Cas Number : 173841-02-6
  • Molecular Weight : 160.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-chloro-1-methyl-1h-pyrazole-5-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6869
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart