ADV412110681 AChemBlock

9-Boc-3-oxa-9-azaspiro[5.5]undecane

  • MDL number MFCD18325172
  • Cas Number : 1259489-90-1
  • Molecular Weight : 255.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/9-boc-3-oxa-9-azaspiro-5-5-undecane.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6871
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart