ADV412110694 AChemBlock

2-Boc-2-azabicyclo[2.2.2]octane-6-carboxylic acid

  • MDL number MFCD18089871
  • Cas Number : 1250997-05-7
  • Molecular Weight : 255.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-boc-2-azabicyclo-2-2-2-octane-6-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6887
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart