ADV412110698 AChemBlock

(3R,4S)-rel-1-Boc-3,4-diaminopyrrolidine

  • MDL number MFCD16876005
  • Cas Number : 945217-60-7
  • Molecular Weight : 201.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3r-4s-rel-1-boc-3-4-diaminopyrrolidine.html
Availability details for
Quantity
  • Availability
  • Add to Cart