ADV412110706 AChemBlock

(S)-N-Boc-2-aminomethylmorpholine Oxalate

  • MDL number MFCD22194319
  • Cas Number : 1373255-08-3
  • Molecular Weight : 306.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form white solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/s-n-boc-2-aminomethylmorpholine-oxalate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=B-1050-01
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart