ADV412110713 AChemBlock

2-Chloro-5-methyl-5H-pyrrolo[3,2-d]pyrimidin-4(3H)-one

  • MDL number MFCD17018620
  • Cas Number : 129872-84-0
  • Molecular Weight : 183.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-5-methyl-5h-pyrrolo-3-2-d-pyrimidin-4-3h-one.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6906
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart