ADV412110722 AChemBlock

Benzyl 5,6-dihydropyridine-1(2H)-carboxylate

  • MDL number MFCD07366998
  • Cas Number : 66207-23-6
  • Molecular Weight : 217.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/benzyl-5-6-dihydropyridine-1-2h-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6915
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart