ADV412110731 AChemBlock

(1R,4R)-2-Methyl-2,5-diazabicyclo[2.2.1]heptane dihydrobromide

  • MDL number MFCD13195528
  • Cas Number : 125224-64-8
  • Molecular Weight : 274

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form pink solid/white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1r-4r-2-methyl-2-5-diazabicyclo-2-2-1-heptane-dihydrobromide.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4130
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart