ADV412110733 AChemBlock

1-Benzhydrylazetidin-3-yl 3-amino-3-iminopropanoate acetate

  • MDL number MFCD09032928
  • Cas Number : 170749-59-4
  • Molecular Weight : 383.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-benzhydrylazetidin-3-yl-3-amino-3-iminopropanoate-acetate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6926
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart